missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Cyclin E (Ser399) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA564734
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-Cyclin E (Ser399) Polyclonal Antibody detects endogenous levels of Cyclin E only when phosphorylated at Ser399.
Phospho-Cyclin E (Ser399) Polyclonal antibody specifically detects Phospho-Cyclin E (Ser399) in Human samples. It is validated for Western Blot.Specifica
Phospho-Cyclin E (Ser399) | |
Polyclonal | |
Unconjugated | |
CCNE1 | |
AW538188; Ccne; Ccne1; CycE1; CYCLE; cyclin E; cyclin E1; cyclin Es; cyclin Et; E-S; G1/S-specific cyclin-E1; pCCNE1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
898 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P24864 | |
CCNE1 | |
A synthesized peptide derived from human Cyclin E1 (Phospho-Ser399) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |