missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ERK1/ERK2 (Thr202, Tyr204) Recombinant Rabbit Monoclonal Antibody (ERK12T202Y204-A11), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537215
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-ERK1/ERK2 (Thr202, Tyr204) Recombinant Monoclonal antibody specifically detects Phospho-ERK1/ERK2 (Thr202, Tyr204) in Human, Mouse samples. It is validated for Flow Cytometry.Specifica
Phospho-ERK1/ERK2 (Thr202, Tyr204) | |
Monoclonal | |
PE | |
MAPK1 | |
12559; 9030612K14Rik; AA407128; AU018647; BcDNA:RE08694; C78273; CG12559; CG12559-PA; CG12559-PC; CG12559-PD; CG12559-PE; CG12559-PG; CG12559-PH; CG12559-PI; CG18732; CT34260; CT39192; DERK; D-ERK; DERK-A; Diphospho-ERK; dm-dpERK; Dmel\CG12559; Dmel_CG12559; DmErk; DmERKA; DmERK-A; DmMAPK; dpERK; dp-ERK; dpERK1; dpMAPK; drosophila ERK; Dsor2; E(sina)7; EC2-1; EK2-1; enhancer of seven in absentia 7; ERK; Erk MAP kinase; Erk/Map kinase; ERK/rolled; Erk1; Erk-1; erk1 erk2; Erk1/2; ERK1b; ERK2; ERK-2; ERKA; ERK-A; ERT1; ERT2; Esrk1; extracellular signal-regulated kinase; extracellular signal-regulated kinase (ERK2); extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase A; extracellular signal-regulated kinase-1; extracellular signal-regulated kinase-2; extracellular signal-related kinase 1; Extracellular-regulated kinase A; extracellular-signal-regulated kinase 2; Extracellular-signal-related kinase A; EY2-2; fi06b09; GroupI | |
Rabbit | |
Protein A/G | |
RUO | |
26413, 26417, 5594, 5595 | |
4° C | |
Liquid |
Flow Cytometry | |
ERK12T202Y204-A11 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P27361, P28482, P63085, Q63844 | |
MAPK1, MAPK3 | |
A synthetic phospho-peptide corresponding to residues surrounding Thr202/Tyr204 of human phospho Erk1/2. | |
10 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |