missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Histone H2A.X (Ser139) Recombinant Rabbit Monoclonal Antibody (HisH2AXS139-1B3), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537024
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-Histone H2A.X (Ser139) Recombinant Monoclonal antibody specifically detects Phospho-Histone H2A.X (Ser139) in Human, Mouse samples. It is validated for Flow Cytometry.Specifica
Phospho-Histone H2A.X (Ser139) | |
Monoclonal | |
PE | |
H2AX | |
AW228881; gamma H2AX; gammaH2ax; H2A histone family member X; H2A histone family, member X; H2A.X; H2A.X variant histone; H2A/X; h2afx; H2ax; H2AX histone; Hist5-2ax; histone 5 protein 2ax; histone H2A.x; Histone H2a/x; Histone H2AX; RGD1566119; similar to H2A histone family, member X; zgc:56329 | |
Rabbit | |
Protein A/G | |
RUO | |
15270, 3014 | |
4° C | |
Liquid |
Flow Cytometry | |
HisH2AXS139-1B3 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P16104, P27661 | |
H2AX | |
A synthetic phospho-peptide corresponding to residues surrounding Ser139 of human phospho histone H2A.X. | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |