missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Lamin A/C (Ser22) Recombinant Rabbit Monoclonal Antibody (LaminACS22-CF12), APC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537184
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-Lamin A/C (Ser22) Recombinant Monoclonal antibody specifically detects Phospho-Lamin A/C (Ser22) in Human, Mouse samples. It is validated for Flow Cytometry.Specifica
Phospho-Lamin A/C (Ser22) | |
Monoclonal | |
APC | |
LMNA | |
70 kDa lamin; cb948; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; I79_009616; IDC; lamin; lamin A; lamin A/C; lamin A/C L homeolog; lamin A/C-like 1; Lamin A+C mutant; Lamin AC; lamin a-c; lamin C; lamin C2; lamin-A; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; lmna; LMNA protein; lmna.L; lmna-A; LMNC; LMNL1; mutant 453W; Mutant lamin A/C; nuclear intermediate filament protein; nuclear lamin A; prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32; RP11-54H19.1; unnamed protein product; wu:fk66d12; XELAEV_18040808mg | |
Rabbit | |
Protein A/G | |
RUO | |
16905, 4000 | |
4° C | |
Liquid |
Flow Cytometry | |
LaminACS22-CF12 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P02545, P48678 | |
LMNA | |
A synthetic phospho-peptide corresponding to residues surrounding Ser22 of human phospho Lamin A/C. | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |