missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MEK3/MEK6 (Ser189, Ser207) Recombinant Rabbit Monoclonal Antibody (MKK3S189MKK6S207-D3), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537055
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-MEK3/MEK6 (Ser189, Ser207) Recombinant Monoclonal antibody specifically detects Phospho-MEK3/MEK6 (Ser189, Ser207) in Human, Mouse samples. It is validated for Flow Cytometry.Specifica
Phospho-MEK3/MEK6 (Ser189, Ser207) | |
Monoclonal | |
PE | |
MAP2K3 | |
AW212142; dual specificity mitogen-activated protein kinase kinase 3; Dual specificity mitogen-activated protein kinase kinase 6; MAP kinase kinase 3; MAP kinase kinase 6; Map2k3; Map2k6; MAPK/ERK kinase 3; MAPK/ERK kinase 6; MAPKK 3; MAPKK 6; MAPKK3; MAPKK6; MEK 3; MEK 6; MEK3; MEK6; mitogen activated protein kinase kinase 3; mitogen activated protein kinase kinase 6; mitogen-activated protein kinase kinase 3; mitogen-activated protein kinase kinase 6; MKK3; MKK6; mMKK3b; OTTHUMP00000166044; Prkmk3; Prkmk6; protein kinase, mitogen activated, kinase 6; protein kinase, mitogen-activated, kinase 3; protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6); SAPK kinase 2; SAPK kinase 3; SAPKK2; SAPKK-2; SAPKK3; SAPKK-3; SKK2; SKK3; Stress-activated protein kinase kinase 2; stress-activated protein kinase kinase 3 | |
Rabbit | |
Protein A/G | |
RUO | |
26397, 26399, 5606, 5608 | |
4° C | |
Liquid |
Flow Cytometry | |
MKK3S189MKK6S207-D3 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
O09110, P46734, P52564, P70236 | |
MAP2K3, MAP2K6 | |
A synthetic phospho-peptide corresponding to residues surrounding Ser189 of human phospho MKK3 and Ser207 of human phospho MKK6. | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |