missing translation for 'onlineSavingsMsg'
Learn More

Phospho-NFkB p65 (Ser529) Rabbit anti-Human, Mouse, Clone: NFkBp65S529-H3, Invitrogen™

Rabbit Monoclonal Antibody

Marca:  Thermo Scientific MA537164

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17106758

  • 552.00€ / 200µL

Spedizione stimata: 16-01-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Phospho-NFkB p65 (Ser529) Monoclonal antibody specifically detects Phospho-NFkB p65 (Ser529) in Human, Mouse samples. It is validated for Flow Cytometry
Specifica

Specifica

Phospho-NFkB p65 (Ser529)
Monoclonal
0.5 mg/mL
PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide
Q04206, Q04207
Rabbit
Protein A/G
RUO
19697, 5970
-20°C
Liquid
Flow Cytometry
NFkBp65S529-H3
Unconjugated
RELA
avian reticuloendotheliosis viral (v-rel) oncogene homolog A; EBP-1; I79_021812; MGC131774; NF KB; NFkappaB p65; nf-kappa-b p65; NF-kappa-B p65delta3; NF-kappa-B transcription factor p65; NF-kappaB transcription factor p65 subunit; NFkB; NF-kB p65 subunit; nfkb rela; NFKB1; nfkb3; NFkB-p50; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit p65; nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; p65; p65 NF kappaB; p65 NF-kappa B; p65 NFkB; P65 transcription factor; putative transcription factor p65 homolog; REL A; rel1; rela; RELA proto-oncogene, NF-kB subunit; rela.L; rela-a; transcription factor p65; v-rel avian reticuloendotheliosis viral oncogene homolog A; v-rel avian reticuloendotheliosis viral oncogene homolog A L homeolog; v-rel reticuloendotheliosis viral oncogene homolog A; v-rel reticuloendotheliosis viral oncogene homolog A (avian); v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappa light polypeptide gene enhancer in B-cells 3, p65; XELAEV_18022301mg; Xrel1; xrela
A synthetic phospho-peptide corresponding to residues surrounding Ser529 of human phospho-NFKB p65.
200 μL
Primary
Mouse, Human
Antibody
IgG1 κ
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali