missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-NFKB p65 (Ser536) Recombinant Rabbit Monoclonal Antibody (NFKBp65S536-B7), APC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537405
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-NFKB p65 (Ser536) Recombinant Monoclonal antibody specifically detects Phospho-NFKB p65 (Ser536) in Human samples. It is validated for Flow Cytometry.Specifica
Phospho-NFKB p65 (Ser536) | |
Monoclonal | |
APC | |
RELA | |
avian reticuloendotheliosis viral (v-rel) oncogene homolog A; EBP-1; I79_021812; MGC131774; NF KB; NFkappaB p65; nf-kappa-b p65; NF-kappa-B p65delta3; NF-kappa-B transcription factor p65; NF-kappaB transcription factor p65 subunit; NFkB; NF-kB p65 subunit; nfkb rela; NFKB1; nfkb3; NFkB-p50; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit p65; nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; p65; p65 NF kappaB; p65 NF-kappa B; p65 NFkB; P65 transcription factor; putative transcription factor p65 homolog; REL A; rel1; rela; RELA proto-oncogene, NF-kB subunit; rela.L; rela-a; Transcription factor p65; v-rel avian reticuloendotheliosis viral oncogene homolog A; v-rel avian reticuloendotheliosis viral oncogene homolog A L homeolog; v-rel reticuloendotheliosis viral oncogene homolog A; v-rel reticuloendotheliosis viral oncogene homolog A (avian); v-rel reticuloendotheliosis viral oncogene homolog A, nuclear fa | |
Rabbit | |
Protein A/G | |
RUO | |
5970 | |
4° C | |
Liquid |
Flow Cytometry | |
NFKBp65S536-B7 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
Q04206 | |
RELA | |
A synthetic phospho-peptide corresponding to residues surrounding Ser536 of human phospho-NFKB p65. | |
10 Tests | |
Primary | |
Human | |
Antibody | |
IgG κ |