missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PKC alpha (Thr497) Recombinant Rabbit Monoclonal Antibody (PKCaT497-F1), FITC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537364
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-PKC alpha (Thr497) Recombinant Monoclonal antibody specifically detects Phospho-PKC alpha (Thr497) in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifica
Phospho-PKC alpha (Thr497) | |
Monoclonal | |
FITC | |
PRKCA | |
AAG6; aging-associated gene 6; AI875142; EC 2.7.11.13; FHC; FTH; FTHL6; im:7139045; kinase PKC-alpha; KPCA; MGC104426; PIG15; PKC III; Pkca; PKC-A; PKC-alpha; PKC-III; PKRCA; PLIF; PRKACA; Prkca; prkcaa; protein kinase C; protein kinase C alpha; protein kinase C alpha type; protein kinase C alpha type; protein kinase C, alpha, a; protein kinase C, alpha; protein kinase C, alpha, a; protein kinase c-alpha; si:dkey-8l19.1; Unknown (protein for IMAGE:7139045); unnamed protein product | |
Rabbit | |
Protein A/G | |
RUO | |
18750, 24680, 5578 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
PKCaT497-F1 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P05696, P17252, P20444 | |
PRKCA | |
A synthetic phospho-peptide corresponding to residues surrounding Thr497 of human phospho PKC alpha. | |
10 Tests | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG κ |