missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PKC gamma (Thr514) Recombinant Rabbit Monoclonal Antibody (PKCgT514-PF4), APC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537195
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-PKC gamma (Thr514) Recombinant Monoclonal antibody specifically detects Phospho-PKC gamma (Thr514) in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifica
Phospho-PKC gamma (Thr514) | |
Monoclonal | |
APC | |
PRKCG | |
PKC; Pkcc; PKCG; PKCgamma; PKC-gamma; PKCI; Prkc; Prkcc; PRKCG; protein kinase C gamma; protein kinase C gamma type; protein kinase C type I (gamma type); protein kinase C, gamma; RATPKCI; SCA14 | |
Rabbit | |
Protein A/G | |
RUO | |
18752, 24681, 5582 | |
4° C | |
Liquid |
Flow Cytometry | |
PKCgT514-PF4 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P05129, P63318, P63319 | |
PRKCG | |
A synthetic phospho-peptide corresponding to residues surrounding Thr514 of human phospho PKC gamma. | |
100 Tests | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG κ |