missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PTEN (Ser380, Thr382, Thr383) Recombinant Rabbit Monoclonal Antibody (PTENS380T382383-E4), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA536994
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-PTEN (Ser380, Thr382, Thr383) Recombinant Monoclonal antibody specifically detects Phospho-PTEN (Ser380, Thr382, Thr383) in Human, Mouse samples. It is validated for Flow Cytometry.Specifica
Phospho-PTEN (Ser380, Thr382, Thr383) | |
Monoclonal | |
PE | |
PTEN | |
10q23del; 2310035O07Rik; A130070J02Rik; AI463227; B430203M17Rik; BZS; CWS1; DEC; GLM2; homolog of human mutated in multiple advanced cancers; MHAM; mitochondrial phosphatase and tensin protein alpha; mitochondrial PTENalpha; Mmac; MMAC1; MMAC1 phosphatase and tensin homolog deleted on chromosome 10; mutated in multiple advanced cancers 1; Phosphatase and tensin homolog; phosphatase and tensin homolog (mutated in multiple advanced cancers 1); phosphatase and tensin homolog deleted on chromosome ten; phosphatase and tensin-like protein; phosphatidylinositol 3; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; phosphatidylinositol-3; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; protein tyrosine phosphatase and tensin-like protein; PTEN; pten tumor suppressor; PTEN1; TEP1 | |
Rabbit | |
Protein A/G | |
RUO | |
19211, 5728 | |
4° C | |
Liquid |
Flow Cytometry | |
PTENS380T382383-E4 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
O08586, P60484 | |
PTEN | |
A synthetic phospho-peptide corresponding to residues surrounding Ser380 and Thr382/383 of human phospho PTEN. | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |