Learn More
Phospho-RPS6 (Ser240/244) Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 29220-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
Ribosomal protein S6 (RPS6), also named as Phosphoprotein NP33, may play an important role in controlling cell growth and proliferation through the selective translation of particular classes of mRNA. Ribosomal protein S6 is the major substrate of protein kinases in eukaryote ribosomes. The phosphorylation is stimulated by growth factors, tumor promoting agents, and mitogens and is dephosphorylated at growth arrest.
Specifica
Phospho-RPS6 (Ser240/244) | |
Polyclonal | |
Unconjugated | |
20 μL | |
6194 | |
Liquid |
Western Blot, ELISA | |
800 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |