missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Shp2 (Tyr580) Rabbit anti-Human, Mouse, APC, Clone: Shp2Y580-4A2, Invitrogen™

Rabbit Monoclonal Antibody

Marca:  Thermo Scientific MA537264

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17106858

  • 165.00€ / pezzi 10

Spedizione stimata: 28-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Phospho-Shp2 (Tyr580) Monoclonal antibody specifically detects Phospho-Shp2 (Tyr580) in Human, Mouse samples. It is validated for Flow Cytometry
Specifica

Specifica

Phospho-Shp2 (Tyr580)
Monoclonal
APC
PTPN11
2700084A17Rik; AW536184; BPTP3; cfc; cSH-PTP2; encodes catalytic domain; HCP; HGNC:9644; JMML; METCDS; MGC14433; Noonan syndrome 1; ns1; null; OTTHUMP00000166108; phosphotyrosyl-protein phosphatase; protein tyrosine phosphatase SH-PTP2; protein tyrosine phosphatase, non-receptor type 11; protein tyrosine phosphatase, non-receptor type 11 (Noonan syndrome 1); protein tyrosine phosphatase, non-receptor type 11 S homeolog; protein-tyrosine phosphatase 1D; Protein-tyrosine phosphatase 2C; protein-tyrosine phosphatase SYP; PTP1C; PTP1D; PTP-1D; ptp-2; ptp2c; PTP-2C; ptpn11; ptpn11.S; ptpn11-a; ptpn11-b; SAP-2; SH2 domain-containing protein tyrosine phosphatase-2; shp2; SHP-2; SHPTP2; SH-PTP2; SH-PTP2 protein tyrosine phosphatase non-receptor type 11; SH-PTP2 protein tyrosine phosphatase, non-receptor type 11; SH-PTP3; Syp; Tyrosine-protein phosphatase non-receptor type 11; XELAEV_18010676mg
A synthetic phospho-peptide corresponding to residues surrounding Tyr580 of human phospho Shp2.
10 Tests
Primary
Mouse, Human
Antibody
IgG1 κ
Flow Cytometry
Shp2Y580-4A2
PBS with 0.20% BSA and 0.09% sodium azide
P35235, Q06124
Rabbit
Protein A/G
RUO
19247, 5781
2-8°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali