missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Stat1 (Tyr701) Rabbit anti-Human, Mouse, Clone: Stat1Y701-3E6, Invitrogen™

Rabbit Monoclonal Antibody

Marca:  Thermo Scientific MA537304

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17106898

  • 275.00€ / 20µL

Spedizione stimata: 07-11-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Phospho-Stat1 (Tyr701) Monoclonal antibody specifically detects Phospho-Stat1 (Tyr701) in Human, Mouse samples. It is validated for Flow Cytometry
Specifica

Specifica

Phospho-Stat1 (Tyr701)
Monoclonal
0.5 mg/mL
PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide
P42224, P42225
Rabbit
Protein A/G
RUO
20846, 6772
-20°C
Liquid
Flow Cytometry
Stat1Y701-3E6
Unconjugated
STAT1
2010005J02Rik; AA408197; CANDF7; DD6G4-4; DKFZp686B04100; IMD31A; IMD31B; IMD31C; ISGF-3; OTTHUMP00000165046; OTTHUMP00000205845; Signal transducer and activator of transcription 1; signal transducer and activator of transcription 1, 91kD; signal transducer and activator of transcription 1, 91kDa; signal transducer and activator of transcription 1-alpha/beta; signal transducer and activator of transcription 1-like; signal transducer and activator of transcription-1; STAT; STAT 1; STAT1; STAT-1; stat1 alpha; STAT91; transcription factor ISGF-3; transcription factor ISGF-3 components p91/p84
A synthetic phospho-peptide corresponding to residues surrounding Tyr701 of human phospho Stat1.
20 μL
Primary
Mouse, Human
Antibody
IgG1 κ
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali