Learn More
Sartorius Pipettatori elettronici PICUS™ a 8 canali
Riduce il rischio di lesioni da sforzi ripetitivi. La pipetta elettronica Sartorius PICUS™ a 8 canali garantisce risultati accurati, precisi e affidabili grazie al motore CC avanzato e al controllo degli errori incorporato.
Marca: Sartorius 735321
1472.41 EUR valido fino al 2024-12-31
Usa il codice promo "21923" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
Light and small electronic pipet ensures an optimal working posture and offers comfortable pipeting. New ergonomic design protects from RSI (repetitive strain injury) and eases workload in long pipetting series. Provides more reliable results with a new generation technology and error control system.
- Helps prevent repetitive strain injury (RSI) due to lightweight, ergonomic design and electronic tip ejection
- Ensures the most accurate, precise and reliable results due to advanced DC-motor concept and built-in error control
- Saves time with extremely fast one-hand adjustment, a unique built-in microplate tracker and easy programmability
- Adjustment wheel for scrolling menus, adjusting volume and controlling manual pipetting
- Variety of pipetting modes in user friendly interface including memory places for favorite programs
- Unique Tracker in conjunction with pipetting, reverse pipetting and multi dispensing modes to help microplate work
- Adjustment possible for 1-point, 2-points and 3-points
- Easy maintenance of lower parts with GLP information integrated in software for service and calibration
- Safe-Cone filters in all models >10 μL to prevent contamination
- Autoclavable lower parts in all models (excluding 1200 μL model)
- Compatible with electronic pipet charging systems, charging stand for one pipet and charging carousel for four pipets
- Direct charging option with micro USB cable
- Delivered without USB AC adapter (available separately)
Specifica
0.9%, 1.5%, 4% | |
Autoclavable | |
8 | |
Yes | |
Da 0.2 a 10 μl | |
0.02 μl | |
Ciascuno |
Puntale da 10 μl | |
Grigio | |
PICUS™ | |
Electronic Pipette | |
Ergonomico | |
Da 3.00 a 0.50 | |
2 anni |