Learn More
PIPES lysis buffer with Triton™ X-100, Thermo Scientific™
Click to view available options
Quantità:
250 mL
500 mL
Dimensione della confezione:
250mL
500mL
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Specifica
| Materiale o nome chimico | PIPES lysis buffer with Triton™ X-100 |
| Forma fisica | Liquid |
| Quantità | 500 mL |