Learn More
Smeg™ Pipette Washing Trolley with Drying System
Used for washing and drying of 100 pipettes with capacity of 1 to 20mL, height up to 450mm. Smeg Instruments Pipette Washing Trolley with Drying System allows fast loading and unloading access.
Marca: Smeg™ LPT100DS
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Made of stainless steel
- Pipette washing disc is fitted in trolley
Specifica
Pipette Washing Trolley with Drying System |