missing translation for 'onlineSavingsMsg'
Learn More

PKLR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17580-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897504

  • 140.00€ / 20µL
Spedizione stimata: 21-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is a pyruvate kinase that catalyzes the transphosphorylation of phohsphoenolpyruvate into pyruvate and ATP, which is the rate-limiting step of glycolysis. Defects in this enzyme, due to gene mutations or genetic variations, are the common cause of chronic hereditary nonspherocytic hemolytic anemia (CNSHA or HNSHA). Multiple transcript variants encoding different isoforms have been found for this gene.
Specifica

Specifica

PKLR
Polyclonal
Unconjugated
PKLR
PK1, PKL, PKLR, PKR, PKRL, Pyruvate kinase 1, Pyruvate kinase isozymes R/L, pyruvate kinase, liver and RBC, R type/L type pyruvate kinase, Red cell/liver pyruvate kinase, RPK
Rabbit
Antigen Affinity Chromatography
RUO
18770, 24651, 5313
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P12928, P30613, P53657
Pklr
PKLR Fusion Protein Ag11096
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali