missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKR Antibody (S04-3F2), Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-19538-100ul
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PKR Monoclonal antibody specifically detects PKR in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifica
| PKR | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| S04-3F2 | |
| Unconjugated | |
| double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated | |
| Recombinant protein of mouse PKR (Uniprot # Q03963) | |
| 100 μg | |
| Apoptosis, Phospho Specific, Signal Transduction | |
| 5610 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto