Learn More
BTX™ Platinum Electrode Wand
Platinum Electrode Wand for delicate and/or difficult tissue transfection.
Marca: BTX™ 450492
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- Simple and easy to use
- Compatible with BTX™ Systems such as Gemini X2 and ECM™ 2001+
Specifica
Ex Vivo Electroporation | |
Platinum | |
Electroporation accessory | |
Platinum Electrode Wand Only, 7 mm, positive | |
7 mm | |
10 μsec. to 100 msec. | |
Non-sterile | |
Square Wave |
BTX™ ECM™ 830, ECM™ 2001+, Gemini X2 and Legacy ECM™ 2001 Generators | |
BTX Electroporation | |
100 V | |
Electrode Wand Only (Positive) | |
1 to 99 (depending on voltage) | |
7 mm | |
Banana to Pin Tip |