Learn More
PLOD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA563182
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: FVSLFFQRLL RLHYPQKHMR LFIHNHEQHH KAQVEEFLAQ HGSEYQSVKL VGPEVRMANA DARN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.Specifica
PLOD1 | |
Polyclonal | |
Unconjugated | |
PLOD1 | |
2410042F05Rik; AI854890; AV007929; EDS6; LH; LH1; LLH; lysine hydroxylase; lysyl hydroxlase 1; Lysyl hydroxylase 1; Plod; Plod1; procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase 1; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 1; procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5351 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q02809 | |
PLOD1 | |
Recombinant protein corresponding to Human PLOD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only