Learn More
SI Analytics™ Combinazione di cavi con spina
Codice prodotto. 10326932 Sfoglia Tutto SI Analytics Prodotti
Click to view available options
Lunghezza (sistema metrico) cavo:
1 m
10 m
2 m
5 m
Tipo:
K 1 A
K 10 A
K2X
K5A
L 1 A
L 1 BNC
L 1 N
L 1 NN
L 2 A
Plug Cable Combination
Unit Size:
Pezzo
Product Code. 10326932
Brand: SI Analytics™ 285122457
Only null left
Add to basket
Add to basket
SI Analytics™ Connection Cables and Plugs are available in various combinations of electrode socket, instrument plug and cable length.
  • Coaxial plug for pH, redox, ammonia and sodium combination electrodes, pH and redox single electrodes as well as reference electrodes in Plus series. The L and LB series plugs are compatible. The LB sockets are matching colors with the BlueLine electrodes
  • Electrode plug for reference electrodes from the predecessor series
  • Plug for conductivity measuring cells with temperature sensor and oxygen cells, for older models
TRUSTED_SUSTAINABILITY
Specifications
Tipi di connettore 4 mm Banana Plug
Tipo L 1 N
Include 1m Coaxial Cable, 4mm Banana Plug
Lunghezza (sistema metrico) cavo 1 m