Learn More
SI Analytics™ Combinazione di cavi con spina
Codice prodotto. 10406812 Sfoglia Tutto SI Analytics Prodotti
Click to view available options
Lunghezza (sistema metrico) cavo:
1 m
10 m
2 m
5 m
Tipo:
K 1 A
K 10 A
K2X
K5A
L 1 A
L 1 BNC
L 1 N
L 1 NN
L 2 A
Plug Cable Combination
Dimensione della confezione:
Pezzo
Codice prodotto. 10406812
Marca: SI Analytics™ 285120217
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
SI Analytics™ Connection Cables and Plugs are available in various combinations of electrode socket, instrument plug and cable length.
  • Coaxial plug for pH, redox, ammonia and sodium combination electrodes, pH and redox single electrodes as well as reference electrodes in Plus series. The L and LB series plugs are compatible. The LB sockets are matching colors with the BlueLine electrodes
  • Electrode plug for reference electrodes from the predecessor series
  • Plug for conductivity measuring cells with temperature sensor and oxygen cells, for older models
TRUSTED_SUSTAINABILITY
Specifica
Tipi di connettore DIN
Tipo K 1 A
Include 1m Coaxial Cable, DIN Instrument Plug
Lunghezza (sistema metrico) cavo 1 m