Learn More
SI Analytics™ Combinazione di cavi con spina
Sfoglia Tutto SI Analytics Prodotti
Click to view available options
Lunghezza (sistema metrico) cavo:
1 m
10 m
2 m
5 m
Tipo:
K 1 A
K 10 A
K2X
K5A
L 1 A
L 1 BNC
L 1 N
L 1 NN
L 2 A
Plug Cable Combination
Dimensione della confezione:
Pezzo
Descrizione
- Coaxial plug for pH, redox, ammonia and sodium combination electrodes, pH and redox single electrodes as well as reference electrodes in Plus series. The L and LB series plugs are compatible. The LB sockets are matching colors with the BlueLine electrodes
- Electrode plug for reference electrodes from the predecessor series
- Plug for conductivity measuring cells with temperature sensor and oxygen cells, for older models
Specifica
Specifica
| Tipi di connettore | DIN |
| Tipo | L 2 A |
| Include | 2m Coaxial Cable, DIN Instrument Plug |
| Lunghezza (sistema metrico) cavo | 2 m |