missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMAL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16965-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The function of this protein remains unknown.Specifica
PNMAL1 | |
Polyclonal | |
Unconjugated | |
PNMA8A | |
PNMA like 1, PNMA like protein 1, PNMAL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55228 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86V59 | |
PNMA8A | |
PNMAL1 Fusion Protein Ag10698 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |