Learn More
POLR3E Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15022-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The POLR3E antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human POLR3E. This antibody recognizes human, mouse, rat antigen. The POLR3E antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifica
| POLR3E | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 55718 | |
| IgG |
| Western Blot, Immunoprecipitation, ELISA | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |