Learn More
Fisherbrand™ Polyethylene Sample Bags
For use in the laboratory to carry samples into a sterile environment.
Marca:  Fisherbrand™ 14955181
Codice prodotto. 11954405
  • ONLINE EXCLUSIVE PRICE
    327.00€
    61.83€ / pezzi 500
    Risparmia 265.17€
    81% Sconto
Solo null disponibili
Spedizione stimata: 12-06-2026
Aggiungi al carrello
Descrizione
Descrizione
  • Secure, contaminant free bags to ensure dependable analysis results
  • Made from heavy gauge, virgin polyethylene that meets FDA specifications
  • Can accommodate solid, semi solid and liquid materials
  • Easy to use, insert sample and seal by twirling the bag away outward four times and bending wire ends inward
  • Economical and efficient way to collect, contain and carry samples