Learn More
Fisherbrand™ Puntali per pipette universali in polipropilene
Eliminate sample contamination and prevent sample loss with Fisherbrand™ Polypropylene Universal Pipette Tips. The polypropylene tips come in packs of 350.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.48600kg
Descrizione
- Sterile
- Blue
- For use with Gilson, Eppendorf, Finnpipette, Bio-Hit, Handrop, Nichiryo and Socorex equipment
Specifica
1000 μl | |
Blue | |
Puntali universali per pipette | |
Polypropylene |
Sterile | |
Autoclavable | |
Gilson™ Pipetman™, Eppendorf Pipetter, Thermo Scientific™ Finnpipette™, Sartorius™ Biohit™ Pipettor, Handrop™, Nichiryo™, Socorex™ Pipettor |