Learn More
Greiner Bio-One Micropiastra per colture cellulari da 1536 pozzetti in polistirene con coperchio
Designed with a rounded square profile for flexible working volume combined with reduced wicking and bubbling. Greiner Bio-One™ Polystyrene 1536 Well HiBase Microplates with Lid are ideally suited for top reading instruments, fluorescence measurements and, due to hydrophobic nature of surface, non polar proteins and peptides.
Marca: Greiner Bio-One 782086
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Clear polystyrene construction for optimum visibility
- Can be traced to production through defined LOT number system
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic, non-cytotoxic
- Manufactured under DIN ISO 9001 guidelines
- Alphanumeric well coding
- Compatible with automated systems
- Square wells with rounded corners combines advantages of square well (i.e. flexible working volume of 3μ to 10μ with the advantages of a round well (reduced wicking and bubbling)
Specifica
DIN ISO 9001 | |
Standard | |
32 x 48 | |
Polystyrene | |
3 to 10 μL | |
Tissue Culture | |
Flat Bottom (F-well) | |
Confezione da 40 |
Free of detectable DNase, RNase, human DNA, non-pyrogenic | |
With Lid | |
Nero | |
1536 | |
Sterile | |
Da 3 a 10 μl | |
Sì |