Learn More
DWK Life Sciences™ Porcelain Desiccator Plate
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Diametro (sistema metrico) esterno:
90 mm
140 mm
190 mm
235 mm
280 mm
Unit Size:
Pezzo
Description
The desiccator insert is a sieve plate that separates the desiccator base into two areas - there is a desiccant in the lower part and the substance to be dried is placed in the upper part. The substance to be dried is placed on the sieve plate in an appropriate container for drying.
- The heat-resistant material allows hot samples to be placed into the desiccator
- Made of heat-resistant material
Specifications
Specifications
| Diametro (sistema metrico) esterno | 235 mm |
| Materiale | Porcelain |
| Da utilizzare con (applicazione) | Drying of materials |