missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine IL-8/CXCL8 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Marca: R&D Systems MAB5351-500
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CXCL8/IL-8 Monoclonal specifically detects CXCL8/IL-8 in Porcine samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifica
| CXCL8/IL-8 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3576 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, ELISA | |
| 105105 | |
| Western Blot 1 ug/mL, Neutralization 1-6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| CAA43461 | |
| IL8 | |
| E. coli-derived recombinant porcine IL-8/CXCL8 Ala26-Gln104 Accession # CAA43461 | |
| 500 μg | |
| Primary | |
| Detects porcine IL-8/CXCL8 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha , 12/SDF-1 beta , rhCXCL13, recombinant mouse CXCL1, 2, 6, 9, 10, 12/SDF-1 alpha , rmCXCL13, recombinant rat (rr) CXCL1, rrCXCL3/CINC2 alpha , or rr3/CINC-2 beta is observed. | |
| Pig | |
| Supernatant |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto