missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP4/PPP4C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-99380-50ul
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PP4/PPP4C Polyclonal antibody specifically detects PP4/PPP4C in Human samples. It is validated for Western Blot
Specifica
| PP4/PPP4C | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.1.3.16, PP4C, PP4protein phosphatase X, catalytic subunit, PP-X, PPXPPH3, protein phosphatase 4 (formerly X), catalytic subunit, protein phosphatase 4, catalytic subunit, Protein phosphatase X | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PP4/PPP4C. | |
| 50 μg | |
| Core ESC Like Genes, Protein Phosphatase, Signal Transduction, Stem Cell Markers | |
| 5531 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto