Learn More
PPARGC1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22378-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.Specifica
| PPARGC1B | |
| Polyclonal | |
| Unconjugated | |
| PPARGC1B | |
| ERRL1, PERC, PGC 1 beta, PGC 1(beta), PGC1, PGC1B, PPAR gamma coactivator 1 beta, PPARGC 1 beta, PPARGC1, PPARGC1B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 133522, 170826 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86YN6 | |
| Ppargc1b | |
| PPARGC1B Fusion Protein Ag17566 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |