missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11651-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein is a specific component of the complex that includes pre-mRNA processing factors PRPF3, PRPF4, and PRPF18, as well as U4/U5/U6 tri-snRNP. This protein has been shown to possess PPIase activity and may act as a protein chaperone that mediates the interactions between different proteins inside the spliceosome.Specifica
PPIH | |
Polyclonal | |
Unconjugated | |
PPIH | |
CYP 20, CYP20, CYPH, PPIase H, PPIH, Rotamase H, SnuCyp 20, USA CYP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10465, 366461, 66101 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43447, Q9D868 | |
Ppih | |
PPIH Fusion Protein Ag2253 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |