missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PRF1 (Human) Recombinant Protein
Human PRF1 full-length ORF ( AAH63043.1, 22 a.a. - 555 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005551-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
- Sequence: PCHTAARSECKRSHKFVPGAWLAGEGVDVTSLRRSGSFPVDTQRFLRPDGTCTLCENALQEGTLQRLPLALTNWRAQGSGCQRHVTRAKVSSTEAVARDAARSIRNDWKVGLDVTPKPTSNVHVSVAGSHSQAANFAAQKTHQDQYSFSTDTVECRFYSFHVVHTPPLHPDFKRALGDLPHHFNASTQPAYLRLISNYGTHFIRAVELGGRISALTALRTCELALEGLTDNEVEDCLTVEAQVNIGIHGSISAEAKACEEKKKKHKMTASFHQTYRERHSEVVGGHHTSINDLLFGIQAGPEQYSAWVNSLPGSPGLVDYTLEPLHVLLDSQDPRREALRRALSQYLTDRARWRDCSRPCPPGRQKSPRDPCQCVCHGSAVTTQDCCPRQRGLAQLEVTFIQAWGLWGDWFTATDAYVKLFLGGQELRTSTVWDNNNPIWSVRLDFGDVLLATGGPLRLQVWDQDSGRDDDLLGTCDQAPKSGSHEVRCNLNHGHLKFRYHARCLPHLGGGTCLDYVPQMLLGEPPGNRSGAVW
Specifica
AAH63043.1 | |
perforin 1 (pore forming protein) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PRF1 | |
GST |
5551 | |
Wheat germ expression system | |
10 μg | |
FLH2, HPLH2, MGC65093, P1, PFN1, PFP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |