Learn More
PRLR Mouse anti-Human, Unconjugated, Clone: 2E10G5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67292-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PRLR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRLR. This antibody recognizes human, mouse antigen. The PRLR antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifica
| PRLR | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot, Immunohistochemistry, ELISA | |
| 2E10G5 | |
| Unconjugated | |
| 150 μL | |
| 5618 | |
| IgG1 |