Learn More
Pro-Lab Diagnostics™ Anse di cavo in nichelcromo certificate/calibrate
Available in nickel-plated brass or lightweight aluminum
Marca: Pro-Lab Diagnostics™ PL106/8-1
Weitere Details : Peso : 1.00000kg
Beinhaltet: Protective case and certificate of calibration
Beschreibung
- Individual loop
- Constructed of nickel-plated brass or aluminum
Spezifikation
8 in. | |
20.3 cm | |
Pro-Loops™ inoculating loops |
Nickel-plated Brass | |
Inoculating Loop Holder |