missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Progesterone Receptor Ab-14, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-14, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-1882-S1

Dettagli aggiuntivi : Peso : 0.00190kg

 vedi altre versioni di questo prodotto

Codice prodotto. 12640397

  • 1740.00€ / 500µL
Spedizione stimata: 17-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: PgR636

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal (shared by A- and B-form)

Immunogen: Formalin fixed recombinant full-length A-form of human progesterone receptor

Molecular Weight: 116kDa (triplet, B-form) and 81kDa (singlet, A-form)

Positive Control: Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifica

Specifica

Progesterone Receptor Ab-14
Monoclonal
Unconjugated
Formalin fixed recombinant full length A-form of human progesterone receptor
500 μL
Cancer and Tumor Biology
Human
IgG2a κ
Immunohistochemistry (Paraffin)
PgR636
Mouse
Breast CA
RUO
Primary
Concentrated
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali