Learn More
Proprotein Convertase 1/PCSK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-98925-100ul
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Proprotein Convertase 1/PCSK1 Polyclonal antibody specifically detects Proprotein Convertase 1/PCSK1 in Human samples. It is validated for ELISA
Specifica
| Proprotein Convertase 1/PCSK1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| BMIQ12;NEC1;Neuroendocrine convertase 1;PC1;PC1/3;PC3;SPC3 | |
| Produced in rabbits immunized with purified, recombinant Human Proprotein Convertase 1/PCSK1 (Accession#: NP_000430.3; Met 1-Arg 617) | |
| 100 μg | |
| Golgi Apparatus Markers | |
| 5122 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |