missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proprotein Convertase 1/PCSK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
259.00€ - 391.00€
Specifica
| Antigene | Proprotein Convertase 1/PCSK1 |
|---|---|
| Diluizione | ELISA 1:5000-1:10000 |
| Applicazioni | ELISA |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
| Product Code | Brand | Quantità | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantità | Price | Quantity & Availability | |||||
|
18611366
|
Novus Biologicals
NBP2-98925-100ul |
100 μg |
391.00€
100µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
18611566
|
Novus Biologicals
NBP2-98925-50ul |
50 μg |
259.00€
50µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Description
Proprotein Convertase 1/PCSK1 Polyclonal antibody specifically detects Proprotein Convertase 1/PCSK1 in Human samples. It is validated for ELISASpecifications
| Proprotein Convertase 1/PCSK1 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Golgi Apparatus Markers | |
| 0.2 um filtered solution in PBS | |
| 5122 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BMIQ12;NEC1;Neuroendocrine convertase 1;PC1;PC1/3;PC3;SPC3 | |
| Produced in rabbits immunized with purified, recombinant Human Proprotein Convertase 1/PCSK1 (Accession#: NP_000430.3; Met 1-Arg 617) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title