missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Epredia™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-321-R7

Dettagli aggiuntivi : Peso : 0.01200kg

 vedi altre versioni di questo prodotto

Codice prodotto. 12600997

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.


Descrizione

Descrizione

Prostatic acid phosphatase (PSAP) is one of the two antigenic markers of prostatic carcinoma, the other being prostate specific antigen. It belongs to the kallikrein family of serine proteases and is suggested to act as a hydrolase to split phospharyl choline in semen and as a transferase.

Host Species: Mouse

Clone: PASE/4LJ

Isotype: IgG1

Species Reactivity: Human. Does not react with dog, rabbit, and rat. Others not known.

Epitope: Not determined

Immunogen: Purified prostatic acid phosphatase (PSAP) from human seminal plasma

Molecular Weight: 52kDa

Positive Control: Normal prostate or prostate CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
Specifica

Specifica

Prostate Specific Acid Phosphatase (PSAP) Ab-1
Monoclonal
Unconjugated
Purified prostatic acid phosphatase (PSAP) from human seminal plasma
IVD
Primary
Prediluted
Immunofluorescence, Immunohistochemistry (Paraffin)
PASE/4LJ
Mouse
7 mL
Cancer and Tumor Biology
Human
IgG1
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali