Learn More
Epredia™ Lab Vision™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody
Provide accurate, reproducible results with the Epredia™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-321-P
Dettagli aggiuntivi : Peso : 0.00250kg
Descrizione
Prostatic acid phosphatase (PSAP) is one of the two antigenic markers of prostatic carcinoma, the other being prostate specific antigen. It belongs to the kallikrein family of serine proteases and is suggested to act as a hydrolase to split phospharyl choline in semen and as a transferase.Host Species: Mouse
Clone: PASE/4LJ
Isotype: IgG1
Species Reactivity: Human. Does not react with dog, rabbit, and rat. Others not known.
Epitope: Not determined
Immunogen: Purified prostatic acid phosphatase (PSAP) from human seminal plasma
Molecular Weight: 52kDa
Positive Control: Normal prostate or prostate CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifica
Prostate Specific Acid Phosphatase (PSAP) Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Cancer and Tumor Biology | |
Human | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
PASE/4LJ | |
Unconjugated | |
Purified prostatic acid phosphatase (PSAP) from human seminal plasma | |
IVD | |
Primary | |
Concentrated |