Learn More
Thermo Scientific™ Proteinase K, Tritirachium album limber
Commonly used in molecular biology to digest protein and remove contamination from preparations of nucleic acid. Addition of Proteinase K to nucleic acid preparations rapidly inactivates nucleases that might otherwise degrade the DNA or RNA during purification.
Marca: Thermo Scientific™ J62051.MB
Dettagli aggiuntivi : CAS : 39450-01-6 Peso : 0.00025kg
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Keep cold | |
25 mg | |
Lyophilized Powder |