missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20180-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. Alternative splicing of this gene results in two transcript variants encoding different isoforms.Specifica
PSAT1 | |
Polyclonal | |
Unconjugated | |
PSAT1 | |
EPIP, Phosphoserine aminotransferase, PSA, PSAT, PSAT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107272, 293820, 29968 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99K85, Q9Y617 | |
Psat1 | |
PSAT1 Fusion Protein Ag14044 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |