Learn More
Pseudouridine Mouse, anti-Human, Clone: 1G7C7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 68578-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The Pseudouridine antibody from Proteintech is a mouse monoclonal antibody generated with ptg of na Pseudouridine. This antibody recognizes human antigen. The Pseudouridine antibody has been validated for the following applications: Dot Blot, ELISA analysis.
Specifica
| Pseudouridine | |
| 1G7C7 | |
| 5-ribosyluracil, Pseudouridine, Ψ | |
| 150 μL | |
| Human | |
| 150 kDa | |
| IgG1 |
| ELISA, Dot Blot | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |