missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSG1 (Human) Recombinant Protein
Human PSG1 full-length ORF ( AAH22338.1, 35 a.a. - 426 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005669-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
- Sequence: QVTIEAEPTKVSEGKDVLLLVHNLPQNLTGYIWYKGQMRDLYHYITSYVVDGEIIIYGPAYSGRETAYSNASLLIQNVTREDAGSYTLHIIKGDDGTRGVTGRFTFTLHLETPKPSISSSNLNPRETMEAVSLTCDPETPDASYLWWMNGQSLPMTHSLKLSETNRTLFLLGVTKYTAGPYECEIRNPVSASRSDPVTLNLLPKLPKPYITINNLNPRENKDVLNFTCEPKSENYTYIWWLNGQSLPVSPRVRRPIENRILILPSVTRNETGPYQCEIRDRYGGIRSDPVTLNVLYGPDLPRIYPSFTYYRSGEVLYLSCSADSNPPAQYSWTINEKFQLPGQKLFIRHITTKHSGLYVCSVRNSATGKESSKSMTVEVSGKWIPASLAIGF
Specifica
AAH22338.1 | |
pregnancy specific beta-1-glycoprotein 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSG1 | |
GST |
5669 | |
Wheat germ expression system | |
25 μg | |
B1G1, CD66f, DHFRP2, FLJ90598, FLJ90654, PBG1, PSBG1, PSGGA, PSGIIA, SP1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |