missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMB5 (Human) Recombinant Protein
Human PSMB5 full-length ORF ( AAH57840.1, 1 a.a. - 263 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005693-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
- Sequence: MALASVLERPLPVNQRGFFGLGGRADLLDLGPGSLSDGLSLAAPGWGVPEEPGIEMLHGTTTLAFKFRHGVIVAADSRATAGAYIASQTVKKVIEINPYLLGTMAGGAADCSFWERLLARQCRIYELRNKERISVAAASKLLANMVYQYKGMGLSMGTMICGWDKRGPGLYYVDSEGNRISGATFSVGSGSVYAYGVMDRGYSYDLEVEQAYDLARRAIYQATYRDAYSGGAVNLYHVREDGWIRVSSDNVADLHEKYSGSTP
Specifica
AAH57840.1 | |
proteasome (prosome, macropain) subunit, beta type, 5 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMB5 | |
GST |
5693 | |
Wheat germ expression system | |
10 μg | |
LMPX, MB1, MGC104214, X | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |