Learn More
PTBP1 Mouse anti-Human, Unconjugated, Clone: 3D6D8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67462-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PTBP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTBP1. This antibody recognizes human, mouse, rat antigen. The PTBP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifica
| PTBP1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 3D6D8 | |
| Unconjugated | |
| 20 μL | |
| 5725 | |
| IgG1 |