Learn More
Bohlender™ PTFE Extender with Adaptor for Syringe Filters
For sterile aeration by means of syringe filters.
Marca:  Bohlender™ D610-50
Dettagli aggiuntivi : Peso : 0.10000kg
Codice prodotto. 10534212
  • 21.15€ / Pezzo
Spedizione stimata: 07-06-2024
per visualizzare lo stock disponibile
Description
Description
  • FDA conformity
  • Made of PTFE
  • With female Luer cone for fixing syringe filters
  • Outer diameter of 3.5 mm
Specifications
Specifications
FDA
White
3.5 mm
Syringe Filter
PTFE
With female Luer cone
Sterile aeration
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers