Learn More
Bohlender™ PTFE Extender with Adaptor for Syringe Filters
For sterile aeration by means of syringe filters.
Marca: Bohlender™ D610-50
Dettagli aggiuntivi : Peso : 0.10000kg
Description
- FDA conformity
- Made of PTFE
- With female Luer cone for fixing syringe filters
- Outer diameter of 3.5 mm
Specifications
FDA | |
White | |
3.5 mm | |
Syringe Filter |
PTFE | |
With female Luer cone | |
Sterile aeration |