Learn More
VACUUBRAND™ PTFE Vacuum Tubings with Small Flanges KF
made of antistatic PTfE, flanges made of stainless steel (1.4305)
Marca: VACUUBRAND™ 20686031
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- hose ribbed only on the outside, almost smooth walls on the inside. Therefore no accumula-tion of liquids or particles like in corrugated hoses and high gas conductivity due to reduced turbulence
- PTFE material with excellent chemical resistance
- PTFE material is antistatic according to BS 5958:1991 / EN ISO 8031, < 107Ohm resistiv-ity between hose and flanges to prevent electrostatic charging on the inside or outside. However, the tubing must not be used for grounding of equipment.
Specifica
Vacuum Tubing | |
16 mm | |
1000mm |
PTFE | |
1 Each |