Learn More
HUBER Pump adaptor cpl.
Includes hose connector NW12
Marca: HUBER 19607
Dettagli aggiuntivi : Peso : 0.36000kg
Descrizione
Pump adapter CPL KISS E, CC-E and baths 208B to 225B and K12 to K25Specifica
MPC-E, CC-E and Baths 208 B to 225 B and K12 to K25 | |
Hose Connectors NW12 | |
1 Ea. | |
Pump Set |